Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308353.1 | NP_001295282.1 | 3 | 322 | ||
NM_005855.3 | NP_005846.1 | 2 | 261 | |||
XM_017003152.1 | XP_016858641.1 | 2 | 183 | |||
XM_017003153.1 | XP_016858642.1 | 2 | 491 | |||
XM_017003154.1 | XP_016858643.1 | 2 | 268 | |||
XM_017003155.1 | XP_016858644.1 | 2 | 189 | |||
XM_017003156.1 | XP_016858645.1 | 2 | 209 | |||
GenBank mRNA | AI017108.1 | - | 2 | 261 | ||
AJ001014.1 | - | 2 | 161 | |||
AY265457.1 | - | 2 | 129 | |||
BC000548.2 | - | 2 | 153 | 3163522 | ||
BI548986.1 | - | 3 | 248 | |||
BM978161.1 | - | 2 | 661 | |||
CR542032.1 | - | 2 | 129 | |||
CR542044.1 | - | 2 | 129 |