Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: RAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308353.1 | NP_001295282.1 | 4 | 448 | ||
NM_005855.3 | NP_005846.1 | 3 | 387 | |||
XM_017003152.1 | XP_016858641.1 | 3 | 309 | |||
XM_017003153.1 | XP_016858642.1 | 3 | 617 | |||
XM_017003154.1 | XP_016858643.1 | 3 | 394 | |||
XM_017003156.1 | XP_016858645.1 | 3 | 335 | |||
GenBank mRNA | AI017108.1 | - | 3 | 387 | ||
AJ001014.1 | - | 3 | 287 | |||
AY265457.1 | - | 3 | 255 | |||
BC000548.2 | - | 3 | 279 | 3163522 | ||
BI548986.1 | - | 4 | 374 | |||
BM126103.1 | - | 1 | 532 | |||
BM978161.1 | - | 1 | 535 | |||
CR542032.1 | - | 3 | 255 | |||
CR542044.1 | - | 3 | 255 |