Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HNRNPUL1 SIRNA Assays ›
Gene Symbol:
HNRNPUL1
Gene Name:
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases:
E1B-AP5, E1BAP5, HNRPUL1
Chromosome Location:
Chr.19: 41262476 - 41307783 on Build GRCh38
UniGene ID:
Hs.155218
Species:
Human
siRNA ID
HSS145967
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HNRNPUL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301016.2
NP_001287945.1
15
2467
NM_001321208.1
NP_001308137.1
15
2507
NM_001321211.1
NP_001308140.1
15
2291
NM_007040.5
NP_008971.2
15
2798
NM_144732.4
NP_653333.1
15
2280
XM_005258459.3
XP_005258516.1
15
2631
XM_005258461.4
XP_005258518.1
15
2461
XM_005258463.3
XP_005258520.1
15
2452
XM_005258464.3
XP_005258521.1
15
2267
XM_005258465.4
XP_005258522.1
15
2478
XM_011526392.2
XP_011524694.1
14
2409
XM_011526393.2
XP_011524695.1
15
2553
XM_011526394.2
XP_011524696.1
15
2308
XM_011526395.2
XP_011524697.1
15
2517
GenBank mRNA
AJ007509.1
-
15
2641
AK001229.1
-
6
1064
AK021455.1
-
4
1083
AK022863.1
-
15
2600
AK127057.1
-
14
2288
AK225850.1
-
15
2290
AK290729.1
-
15
2262
AK291315.1
-
16
2547
AK291762.1
-
15
2600
AK297127.1
-
15
2395
AK308156.1
-
16
2310
AL050146.1
-
8
1406
BC002564.2
-
15
2538
3139049
BC004242.1
-
3
442
BC009988.2
-
15
2444
4127873
BC014232.2
-
15
2419
4558792
BC027713.2
-
15
2365
4123077
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA processing
response to virus
RNA binding
protein binding
enzyme binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0