Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DDX19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014449.2 | NP_001014449.1 | 5 | 553 | ||
NM_001014451.2 | NP_001014451.1 | 6 | 563 | |||
NM_001257172.1 | NP_001244101.1 | 6 | 488 | |||
NM_001257173.1 | NP_001244102.1 | 6 | 645 | |||
NM_001257174.1 | NP_001244103.1 | 5 | 599 | |||
NM_001257175.1 | NP_001244104.1 | 4 | 460 | |||
NM_007242.5 | NP_009173.1 | 7 | 656 | |||
XM_006721127.2 | XP_006721190.1 | 6 | 578 | |||
XM_011522832.2 | XP_011521134.1 | 8 | 712 | |||
XM_011522833.2 | XP_011521135.1 | 7 | 619 | |||
XM_011522834.2 | XP_011521136.1 | 7 | 601 | |||
XM_017022890.1 | XP_016878379.1 | 7 | 694 | |||
GenBank mRNA | AF353720.1 | - | 5 | 599 | ||
AJ237946.1 | - | 7 | 517 | |||
AK027378.1 | - | 7 | 621 | |||
AK223115.1 | - | 7 | 627 | |||
AK296868.1 | - | 4 | 457 | |||
AK301938.1 | - | 6 | 645 | |||
AK302107.1 | - | 6 | 488 | |||
AK316346.1 | - | 5 | 534 | |||
AL136639.1 | - | 6 | 532 | |||
BC003626.2 | - | 7 | 620 | 3030038 | ||
BC010008.2 | - | 5 | 505 | 4298615 | ||
BC110805.1 | - | 6 | 427 | |||
BI756088.1 | - | 3 | 314 | |||
CR457215.1 | - | 7 | 517 | |||
CR533509.1 | - | 6 | 424 |