Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PUF60 SIRNA Assays ›
Gene Symbol:
PUF60
Gene Name:
poly(U) binding splicing factor 60
Gene Aliases:
FIR, RoBPI, SIAHBP1, VRJS
Chromosome Location:
Chr.8: 143816344 - 143829386 on Build GRCh38
UniGene ID:
Hs.521924
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS146065
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PUF60
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136033.2
NP_001129505.1
11, 12
1474
NM_001271096.1
NP_001258025.1
10, 11
1401
NM_001271097.1
NP_001258026.1
9, 10
1317
NM_001271098.1
NP_001258027.1
11, 12
1452
NM_001271099.1
NP_001258028.1
10, 11
1368
NM_001271100.1
NP_001258029.1
10, 11
1423
NM_014281.4
NP_055096.2
10, 11
1404
NM_078480.2
NP_510965.1
11, 12
1455
XM_011516929.2
XP_011515231.1
12, 13
1654
XM_011516930.2
XP_011515232.1
11, 12
1505
XM_017013234.1
XP_016868723.1
11, 12
2987
XM_017013235.1
XP_016868724.1
11, 12
1557
XM_017013236.1
XP_016868725.1
10, 11
2936
XM_017013237.1
XP_016868726.1
12, 13
1716
XM_017013238.1
XP_016868727.1
12, 13
1516
XM_017013239.1
XP_016868728.1
11, 12
1482
XM_017013240.1
XP_016868729.1
10, 11
1431
GenBank mRNA
AF114818.1
-
11, 12
1436
AF190744.1
-
11, 12
1364
AF217197.2
-
10, 11
1331
AK292373.1
-
11, 12
1436
BC008875.2
-
10, 11
1370
4138778
BC009734.1
-
10, 11
1353
3877563
BC011265.1
-
11, 12
1391
4155549
BC011979.1
-
9, 10
1270
3863660
BC035435.2
-
10, 11
1355
CR457143.1
-
11, 12
1372
U51586.1
-
9, 10
1318
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
apoptotic process
RNA splicing
cell-cell adhesion
nucleotide binding
DNA binding
protein binding
identical protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-79lxv:80/100.66.77.52:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0