Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NT5C2 SIRNA Assays ›
Gene Symbol:
NT5C2
Gene Name:
5'-nucleotidase, cytosolic II
Gene Aliases:
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
Chromosome Location:
Chr.10: 103088017 - 103193306 on Build GRCh38
UniGene ID:
Hs.97439
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS146102
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NT5C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134373.2
NP_001127845.1
2
142
NM_012229.4
NP_036361.1
3
240
XM_005269632.4
XP_005269689.1
2
127
XM_005269633.4
XP_005269690.1
3
220
XM_005269634.4
XP_005269691.1
3
252
XM_005269635.4
XP_005269692.1
2
442
XM_005269636.4
XP_005269693.1
3
252
XM_005269640.4
XP_005269697.1
3
206
XM_005269641.4
XP_005269698.1
2
108
XM_005269643.4
XP_005269700.1
2
108
XM_005269644.4
XP_005269701.1
3
206
XM_005269645.4
XP_005269702.1
2
108
XM_005269646.4
XP_005269703.1
2
108
XM_006717721.3
XP_006717784.1
3
206
XM_006717723.3
XP_006717786.1
2
108
XM_011539537.2
XP_011537839.1
3
252
XM_017015948.1
XP_016871437.1
3
252
XM_017015949.1
XP_016871438.1
2
108
XM_017015950.1
XP_016871439.1
2
108
XM_017015951.1
XP_016871440.1
2
108
XM_017015952.1
XP_016871441.1
2
108
XM_017015953.1
XP_016871442.1
2
442
XM_017015954.1
XP_016871443.1
2
442
XM_017015955.1
XP_016871444.1
2
108
XM_017015956.1
XP_016871445.1
2
442
XM_017015957.1
XP_016871446.1
2
108
XM_017015958.1
XP_016871447.1
3
206
XM_017015959.1
XP_016871448.1
2
108
XM_017015961.1
XP_016871450.1
2
442
XM_017015962.1
XP_016871451.1
2
108
XM_017015963.1
XP_016871452.1
3
252
XM_017015964.1
XP_016871453.1
3
252
XM_017015965.1
XP_016871454.1
2
129
XM_017015966.1
XP_016871455.1
2
108
XM_017015967.1
XP_016871456.1
2
108
XM_017015968.1
XP_016871457.1
3
206
XM_017015969.1
XP_016871458.1
2
108
XM_017015971.1
XP_016871460.1
2
122
XM_017015972.1
XP_016871461.1
2
108
XM_017015973.1
XP_016871462.1
2
108
XM_017015974.1
XP_016871463.1
3
206
XM_017015975.1
XP_016871464.1
2
108
XM_017015976.1
XP_016871465.1
1
71
GenBank mRNA
AK291667.1
-
2
108
AK312517.1
-
2
108
BC001595.1
-
2
191
3543103
BX648078.1
-
2
188
D38524.1
-
2
130
DA879821.1
-
3
209
DB169780.1
-
2
144
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide catabolic process
phosphorylation
dephosphorylation
drug metabolic process
IMP metabolic process
adenosine metabolic process
nucleotide binding
protein binding
5'-nucleotidase activity
metal ion binding
nucleoside phosphotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0