Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EFR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319099.1 | NP_001306028.1 | 3 | 308 | ||
NM_014971.1 | NP_055786.1 | 3 | 320 | |||
XM_011532701.2 | XP_011531003.1 | 3 | 320 | |||
GenBank mRNA | AB023170.1 | - | 3 | 294 | ||
AK095190.1 | - | 3 | 308 | |||
DA241650.1 | - | 3 | 320 |