Search
Search

SV2C| Gene Symbol: SV2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297716.1 | NP_001284645.1 | 9 | 1593 | ||
| NM_014979.3 | NP_055794.3 | 9 | 1593 | |||
| XM_011543281.2 | XP_011541583.1 | 9 | 1960 | |||
| XM_011543282.2 | XP_011541584.2 | 9 | 2520 | |||
| XM_017009244.1 | XP_016864733.1 | 5 | 511 | |||
| GenBank mRNA | AB028977.1 | - | 7 | 654 | ||
| AK094917.1 | - | 9 | 1594 | |||
| BC100824.1 | - | 9 | 1529 | 40001296 | ||
| BC100825.1 | - | 9 | 1527 | 40001297 | ||
| BC100826.1 | - | 9 | 1527 | 40001302 | ||
| BC100827.1 | - | 9 | 1527 | 40001303 | ||