Search
Search

KIF21B| Gene Symbol: KIF21B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252100.1 | NP_001239029.1 | 33 | 4885 | ||
| NM_001252102.1 | NP_001239031.1 | 33 | 4885 | |||
| NM_001252103.1 | NP_001239032.1 | 32 | 4846 | |||
| NM_017596.3 | NP_060066.2 | 32 | 4846 | |||
| XM_017000731.1 | XP_016856220.1 | 32 | 4528 | |||
| XM_017000732.1 | XP_016856221.1 | 27 | 3669 | |||
| GenBank mRNA | AB007918.2 | - | 32 | 4846 | ||
| AM177180.1 | - | 33 | 4574 | |||
| BC078676.1 | - | 4 | 469 | |||
| BC137281.1 | - | 33 | 4593 | 9021278 | ||
| BC144557.1 | - | 32 | 4554 | 9053085 | ||