Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TNRC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024843.1 | NP_001020014.1 | 23 | 2915 | ||
NM_001162501.1 | NP_001155973.1 | 22 | 5236 | |||
NM_015088.2 | NP_055903.2 | 20 | 4906 | |||
GenBank mRNA | AB029016.2 | - | 20 | 4886 | ||
AK294519.1 | - | 22 | 5227 | |||
AK304087.1 | - | 17 | 2391 | |||
BC028626.2 | - | 23 | 2848 | 4826787 | ||
BX647630.1 | - | 17 | 4108 |