Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZCCHC11 SIRNA Assays ›
Gene Symbol:
ZCCHC11
Gene Name:
zinc finger CCHC-type containing 11
Gene Aliases:
PAPD3, TUT4
Chromosome Location:
Chr.1: 52423275 - 52553463 on Build GRCh38
UniGene ID:
Hs.655407
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS146317
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZCCHC11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001009881.2
NP_001009881.1
6
1465
NM_015269.2
NP_056084.1
6
1465
XM_005270676.2
XP_005270733.1
6
1474
XM_005270678.2
XP_005270735.1
6
1474
XM_005270679.2
XP_005270736.1
6
1474
XM_005270680.2
XP_005270737.1
6
1474
XM_005270681.2
XP_005270738.1
6
1474
XM_005270682.2
XP_005270739.1
6
1474
XM_005270683.2
XP_005270740.1
6
1474
XM_006710498.2
XP_006710561.1
6
1411
XM_006710499.3
XP_006710562.1
6
1418
XM_006710500.1
XP_006710563.1
6
1474
XM_011541097.1
XP_011539399.1
7
1496
XM_011541098.1
XP_011539400.1
6
1474
XM_011541099.1
XP_011539401.1
6
1474
XM_011541100.2
XP_011539402.1
6
1474
XM_011541101.2
XP_011539403.1
6
1474
XM_017000803.1
XP_016856292.1
6
1474
XM_017000804.1
XP_016856293.1
6
1474
XM_017000805.1
XP_016856294.1
6
1474
XM_017000806.1
XP_016856295.1
6
1474
XM_017000807.1
XP_016856296.1
6
1474
XM_017000808.1
XP_016856297.1
6
1474
XM_017000809.1
XP_016856298.1
6
1474
XM_017000810.1
XP_016856299.1
6
1474
XM_017000811.1
XP_016856300.1
6
1474
XM_017000812.1
XP_016856301.1
6
1474
XM_017000813.1
XP_016856302.1
6
1474
XM_017000814.1
XP_016856303.1
6
1474
XM_017000815.1
XP_016856304.1
6
1474
XM_017000816.1
XP_016856305.1
6
1474
XM_017000817.1
XP_016856306.1
6
1474
XM_017000818.1
XP_016856307.1
6
1474
GenBank mRNA
AK095733.1
-
6
1475
AK307579.1
-
6
1474
AK308427.1
-
6
680
BC131734.1
-
6
1357
40125005
BX648783.1
-
5
862
D83776.1
-
5
830
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotidyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
miRNA metabolic process
miRNA catabolic process
stem cell population maintenance
pre-miRNA processing
RNA 3'-end processing
histone mRNA catabolic process
protein binding
zinc ion binding
poly(A) RNA binding
RNA uridylyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline