Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TENM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098816.2 | NP_001092286.2 | 29, 30 | 5733 | ||
XM_011544933.2 | XP_011543235.1 | 12, 13 | 2828 | |||
XM_017017525.1 | XP_016873014.1 | 28, 29 | 6004 | |||
XM_017017526.1 | XP_016873015.1 | 26, 27 | 6909 | |||
GenBank mRNA | AB037723.3 | - | 14, 15 | 3011 | ||
AK127101.1 | - | 1 | 28 | |||
AK127705.1 | - | 2, 3 | 662 | |||
BX537983.1 | - | 1, 2 | 177 | |||
BX640737.1 | - | 3, 4 | 1160 |