Search
Search

MYLPF| Gene Symbol: MYLPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324458.1 | NP_001311387.1 | 9 | 665 | ||
| NM_001324459.1 | NP_001311388.1 | 9 | 693 | |||
| NM_013292.4 | NP_037424.2 | 7 | 528 | |||
| GenBank mRNA | AA662706.1 | - | 1 | 178 | ||
| AF363061.1 | - | 7 | 501 | |||
| AK314082.1 | - | 7 | 528 | |||
| BC012571.1 | - | 7 | 506 | 4245958 | ||
| BX649054.1 | - | 7 | 496 | |||
| CR456963.1 | - | 6 | 447 | |||
| M21812.1 | - | 7 | 508 | |||
| X57543.1 | - | 7 | 508 | |||