Search
Search

PRRX2| Gene Symbol: PRRX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016307.3 | NP_057391.1 | 3 | 810 | ||
| XM_017014803.1 | XP_016870292.1 | 3 | 418 | |||
| GenBank mRNA | AF061970.1 | - | 3 | 613 | ||
| BC014645.1 | - | 3 | 810 | 4553935 | ||
| U81600.1 | - | 2 | 316 | |||