Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SETD5 SIRNA Assays ›
Gene Symbol:
SETD5
Gene Name:
SET domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 9397700 - 9478154 on Build GRCh38
UniGene ID:
Hs.288164
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS147924
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SETD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080517.2
NP_001073986.1
4
561
NM_001292043.1
NP_001278972.1
5
786
XM_005265301.1
XP_005265358.1
4
542
XM_005265303.1
XP_005265360.1
5
786
XM_011533920.1
XP_011532222.1
6
979
XM_011533921.1
XP_011532223.1
5
581
XM_011533922.1
XP_011532224.1
4
542
XM_011533927.1
XP_011532229.1
6
979
XM_011533928.1
XP_011532230.1
4
542
XM_011533930.1
XP_011532232.1
5
786
XM_011533932.1
XP_011532234.1
5
786
XM_011533933.1
XP_011532235.1
2
114
XM_017006767.1
XP_016862256.1
6
2641
XM_017006768.1
XP_016862257.1
4
2155
XM_017006769.1
XP_016862258.1
5
4297
XM_017006770.1
XP_016862259.1
5
4297
XM_017006771.1
XP_016862260.1
4
542
XM_017006772.1
XP_016862261.1
5
4297
XM_017006773.1
XP_016862262.1
5
4297
XM_017006774.1
XP_016862263.1
4
542
XM_017006775.1
XP_016862264.1
5
4297
XM_017006776.1
XP_016862265.1
5
786
XM_017006777.1
XP_016862266.1
5
786
XM_017006778.1
XP_016862267.1
5
786
XM_017006779.1
XP_016862268.1
5
786
XM_017006780.1
XP_016862269.1
5
786
XM_017006781.1
XP_016862270.1
5
786
XM_017006782.1
XP_016862271.1
5
4297
XM_017006783.1
XP_016862272.1
6
979
XM_017006784.1
XP_016862273.1
5
4297
XM_017006785.1
XP_016862274.1
5
4297
XM_017006786.1
XP_016862275.1
5
4297
GenBank mRNA
AK092046.1
-
5
586
CR627408.1
-
2
138
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_028385, AK164036, BC083184, BC056502, AK122551.
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0