Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMKLB SIRNA Assays ›
Gene Symbol:
RIMKLB
Gene Name:
ribosomal modification protein rimK like family member B
Gene Aliases:
FAM80B, NAAGS, NAAGS-I
Chromosome Location:
Chr.12: 8681606 - 8783098 on Build GRCh38
UniGene ID:
Hs.504670
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS148526
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RIMKLB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001297776.1
NP_001284705.1
5
1134
NM_020734.3
NP_065785.2
5
1566
NR_123740.1
5
878
XM_006719118.2
XP_006719181.1
5
1255
XM_006719127.2
XP_006719190.1
5
1255
XM_011520776.1
XP_011519078.1
5
1255
XM_011520777.1
XP_011519079.1
5
1255
XM_011520778.2
XP_011519080.1
5
1255
XM_011520779.2
XP_011519081.1
5
1255
XM_011520780.1
XP_011519082.1
5
1255
XM_011520783.1
XP_011519085.1
5
1255
XM_011520790.1
XP_011519092.1
6
1942
XM_017019677.1
XP_016875166.1
5
1255
XM_017019678.1
XP_016875167.1
5
1255
XM_017019679.1
XP_016875168.1
5
1255
XM_017019680.1
XP_016875169.1
5
1255
XM_017019681.1
XP_016875170.1
5
1255
XM_017019682.1
XP_016875171.1
5
1134
XM_017019683.1
XP_016875172.1
6
1942
XM_017019684.1
XP_016875173.1
5
878
XM_017019685.1
XP_016875174.1
6
1657
XM_017019686.1
XP_016875175.1
5
892
XM_017019687.1
XP_016875176.1
6
1256
XM_017019688.1
XP_016875177.1
5
1396
XM_017019689.1
XP_016875178.1
5
945
XM_017019690.1
XP_016875179.1
5
756
XM_017019691.1
XP_016875180.1
5
1134
XM_017019692.1
XP_016875181.1
5
1112
XM_017019693.1
XP_016875182.1
5
1134
XM_017019694.1
XP_016875183.1
5
1139
XM_017019695.1
XP_016875184.1
5
908
XM_017019696.1
XP_016875185.1
6
1942
XM_017019697.1
XP_016875186.1
5
829
GenBank mRNA
AB033064.1
-
5
1505
AK302847.1
-
5
1134
BC006368.2
-
6
1827
BC021977.2
-
2
230
BC033793.1
-
5
875
5139256
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
ATP binding
metal ion binding
N-acetyl-L-aspartate-L-glutamate ligase activity
citrate-L-glutamate ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0