Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACBD4 SIRNA Assays ›
Gene Symbol:
ACBD4
Gene Name:
acyl-CoA binding domain containing 4
Gene Aliases:
HMFT0700
Chromosome Location:
Chr.17: 45132600 - 45144181 on Build GRCh38
UniGene ID:
Hs.110298
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS149264
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ACBD4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135705.2
NP_001129177.1
10
1226
NM_001135706.2
NP_001129178.1
10
1264
NM_001135707.2
NP_001129179.1
10
2076
NM_001321352.1
NP_001308281.1
12
1388
NM_001321353.1
NP_001308282.1
12
1385
NM_024722.3
NP_078998.1
9
1380
XM_006722085.2
XP_006722148.1
11
1599
XM_017025084.1
XP_016880573.1
11
1655
XM_017025085.1
XP_016880574.1
12
1406
XM_017025086.1
XP_016880575.1
13
1555
XM_017025087.1
XP_016880576.1
11
1637
XM_017025088.1
XP_016880577.1
11
1634
XM_017025090.1
XP_016880579.1
11
1617
XM_017025091.1
XP_016880580.1
11
1596
XM_017025092.1
XP_016880581.1
12
1721
XM_017025094.1
XP_016880583.1
9
2244
XM_017025095.1
XP_016880584.1
10
1515
XM_017025096.1
XP_016880585.1
10
1477
XM_017025097.1
XP_016880586.1
10
1494
XM_017025098.1
XP_016880587.1
11
1245
XM_017025099.1
XP_016880588.1
10
1456
GenBank mRNA
AB073387.1
-
10
1176
AK023384.1
-
9
1340
AK296167.1
-
10
1251
AK316359.1
-
10
1264
BC029164.1
-
10
1165
5188183
BC041143.1
-
12
1357
5769682
KU178779.1
-
9
920
KU178780.1
-
9
885
KU178781.1
-
9
882
KU178782.1
-
8
742
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Process(es)
fatty-acyl-CoA binding
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0