Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CYB5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030579.2 | NP_085056.2 | 1, 2 | 264 | ||
GenBank mRNA | AB009282.1 | - | 1, 2 | 291 | ||
AK291576.1 | - | 1, 2 | 196 | |||
BC004373.1 | - | 1, 2 | 199 | 3639133 | ||
BC014431.1 | - | 1, 2 | 222 | 3542294 | ||
BX647922.1 | - | 1, 2 | 240 |