Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L12 SIRNA Assays ›
Gene Symbol:
BCL2L12
BCL2L12P1
Gene Name:
BCL2 like 12
BCL2 like 12 pseudogene 1
Gene Aliases:
-
-
Chromosome Location:
Chr.19: 49665072 - 49673916 on Build GRCh38
Chromosome Location:
Chr.3: 131526468 - 131527141 on Build GRCh38
UniGene ID:
Hs.289052
Hs.640518
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
HSS149675
Size
Availability
Made To Order
Catalog #
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040668.1
NP_001035758.1
1, 2
935
NM_001282516.1
NP_001269445.1
1, 2
935
NM_001282517.1
NP_001269446.1
1, 2
935
NM_001282519.1
NP_001269448.1
1, 2
935
NM_001282520.1
NP_001269449.1
1, 2
935
NM_001282521.1
NP_001269450.1
1, 2
935
NM_138639.1
NP_619580.1
1, 2
935
NR_104200.1
1, 2
935
NR_104201.1
1, 2
935
NR_104202.1
1, 2
935
NR_104203.1
1, 2
935
NR_104204.1
1, 2
935
NR_104205.1
1, 2
935
XM_017027345.1
XP_016882834.1
1, 2
1005
XM_017027346.1
XP_016882835.1
1, 2
1005
GenBank mRNA
BC007724.2
-
1, 2
70
BC104004.1
-
1, 2
511
40024338
BC104005.1
-
1, 2
511
40024342
BC104006.1
-
1, 2
511
40024345
BI159786.1
-
1, 2
101
FJ868798.1
-
1, 2
253
FJ868799.1
-
1, 2
253
FJ868800.1
-
1, 2
253
FJ868801.1
-
1, 2
253
FJ868802.1
-
1, 2
253
FJ868803.1
-
1, 2
253
FJ911906.1
-
1, 2
253
FJ911907.1
-
1, 2
253
FJ911908.1
-
1, 2
253
HM347053.1
-
1, 2
253
KR820244.1
-
1
870
KR820245.1
-
1
870
KR820246.1
-
1
870
KR820247.1
-
1
870
KR820248.1
-
1
870
KR820249.1
-
1
870
KR820250.1
-
1
870
KR820251.1
-
1
870
KR820252.1
-
1
870
KR820253.1
-
1
870
KR820254.1
-
1
870
KR820255.1
-
1
870
KR820256.1
-
1
870
KR820257.1
-
1
870
KR820258.1
-
1
870
KR820259.1
-
1
870
KR820260.1
-
1
870
KR820261.1
-
1
870
KR820262.1
-
1
870
KR820263.1
-
1
870
KR820264.1
-
1
870
KR820265.1
-
1
870
KR820266.1
-
1
870
KR820267.1
-
1
870
KR820268.1
-
1
870
KR820269.1
-
1
870
KR820270.1
-
1, 2
253
KR820271.1
-
1, 2
253
KR820272.1
-
1, 2
253
KR820273.1
-
1, 2
253
KR820274.1
-
1, 2
253
KR820275.1
-
1, 2
253
KR820276.1
-
1, 2
253
KR820277.1
-
1, 2
253
KR820278.1
-
1, 2
253
KR820279.1
-
1, 2
253
KR820280.1
-
1, 2
253
KR820281.1
-
1, 2
253
KR820282.1
-
1, 2
253
KR820283.1
-
1, 2
253
KR820284.1
-
1, 2
253
KR820285.1
-
1, 2
253
KR820286.1
-
1, 2
253
KR820287.1
-
1, 2
253
KR820288.1
-
1
870
KR820289.1
-
1
870
KR820290.1
-
1
870
KR820291.1
-
1
870
KR820292.1
-
1
870
KR820293.1
-
1
870
Gene Symbol: BCL2L12P1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
GQ202621.1
-
1
82
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of cellular senescence
p53 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-kkgv7:80/100.66.73.110:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0