Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OBSCN SIRNA Assays ›
          Gene Symbol:
          OBSCN
          Gene Name:
          obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
          Gene Aliases:
          ARHGEF30, UNC89
          Chromosome Location:
          Chr.1: 228208013 - 228381431 on Build GRCh38
          UniGene ID:
          Hs.650039
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS149790
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: OBSCN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098623.2 NP_001092093.2 60, 61 16149
          NM_001271223.2 NP_001258152.2 71, 72 19020
          NM_052843.3 NP_443075.3 60, 61 16149
          XM_005273287.4 XP_005273344.1 72, 73 19400
          XM_005273291.4 XP_005273348.1 72, 73 19364
          XM_005273298.4 XP_005273355.1 71, 72 19091
          XM_005273307.4 XP_005273364.1 67, 68 18278
          XM_006711822.3 XP_006711885.1 70, 71 18824
          XM_006711823.3 XP_006711886.1 70, 71 18824
          XM_006711827.3 XP_006711890.1 64, 65 17239
          XM_006711829.3 XP_006711892.1 63, 64 16963
          XM_011544297.2 XP_011542599.1 64, 65 17240
          XM_011544299.2 XP_011542601.1 63, 64 16964
          XM_017002443.1 XP_016857932.1 72, 73 19445
          XM_017002444.1 XP_016857933.1 72, 73 19408
          XM_017002445.1 XP_016857934.1 72, 73 19445
          XM_017002446.1 XP_016857935.1 72, 73 19445
          XM_017002447.1 XP_016857936.1 72, 73 19442
          XM_017002448.1 XP_016857937.1 72, 73 19295
          XM_017002449.1 XP_016857938.1 71, 72 19182
          XM_017002450.1 XP_016857939.1 71, 72 19182
          XM_017002451.1 XP_016857940.1 71, 72 19182
          XM_017002452.1 XP_016857941.1 71, 72 19182
          XM_017002453.1 XP_016857942.1 71, 72 19169
          XM_017002454.1 XP_016857943.1 71, 72 19169
          XM_017002455.1 XP_016857944.1 71, 72 19169
          XM_017002456.1 XP_016857945.1 71, 72 19169
          XM_017002457.1 XP_016857946.1 71, 72 19169
          XM_017002458.1 XP_016857947.1 70, 71 18918
          XM_017002459.1 XP_016857948.1 70, 71 18918
          XM_017002460.1 XP_016857949.1 70, 71 18918
          XM_017002461.1 XP_016857950.1 70, 71 18905
          XM_017002462.1 XP_016857951.1 70, 71 18893
          XM_017002463.1 XP_016857952.1 70, 71 18872
          XM_017002464.1 XP_016857953.1 70, 71 18821
          XM_017002465.1 XP_016857954.1 69, 70 18654
          XM_017002466.1 XP_016857955.1 69, 70 18654
          XM_017002467.1 XP_016857956.1 69, 70 18654
          XM_017002468.1 XP_016857957.1 68, 69 18390
          XM_017002469.1 XP_016857958.1 66, 67 17862
          XM_017002470.1 XP_016857959.1 61, 62 16543
          XM_017002471.1 XP_016857960.1 72, 73 19444
          XM_017002472.1 XP_016857961.1 72, 73 19444
          XM_017002473.1 XP_016857962.1 72, 73 19444
          GenBank mRNA AJ002535.1 - 61, 62 16146
          AL832357.1 - 5, 6 565

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          multicellular organism development
          regulation of Rho protein signal transduction
          protein localization to M-band
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          sarcomere organization
          regulation of small GTPase mediated signal transduction
          mitophagy in response to mitochondrial depolarization
          protein serine/threonine kinase activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          protein binding
          calmodulin binding
          ATP binding
          structural constituent of muscle
          ankyrin binding
          titin binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline