Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: C7orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134395.1 | NP_001127867.1 | 5 | 574 | ||
NM_001134396.1 | NP_001127868.1 | 5 | 555 | |||
NM_001318252.1 | NP_001305181.1 | 5 | 1458 | |||
NM_032350.5 | NP_115726.1 | 5 | 584 | |||
NR_134537.1 | 4 | 560 | ||||
XM_006715793.2 | XP_006715856.1 | 4 | 1198 | |||
XM_011515580.2 | XP_011513882.1 | 4 | 1198 | |||
XM_011515581.2 | XP_011513883.1 | 5 | 618 | |||
XM_011515582.2 | XP_011513884.1 | 5 | 569 | |||
XM_011515583.2 | XP_011513885.1 | 5 | 593 | |||
XM_011515584.2 | XP_011513886.1 | 5 | 1274 | |||
XM_017012719.1 | XP_016868208.1 | 4 | 1198 | |||
XM_017012720.1 | XP_016868209.1 | 5 | 578 | |||
XM_017012721.1 | XP_016868210.1 | 6 | 1497 | |||
XM_017012722.1 | XP_016868211.1 | 5 | 617 | |||
GenBank mRNA | AF495721.1 | - | 5 | 1414 | ||
BC006224.2 | - | 5 | 584 | 3941780 | ||
BM762287.1 | - | 4 | 504 | |||
BM763235.1 | - | 5 | 558 |