Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320845.1 | NP_001307774.1 | 4 | 477 | ||
NM_133491.4 | NP_597998.1 | 4 | 460 | |||
XM_017024073.1 | XP_016879562.1 | 4 | 477 | |||
GenBank mRNA | AF348524.1 | - | 4 | 219 | ||
BC011751.2 | - | 4 | 430 | 3534187 | ||
BC065922.1 | - | 1 | 769 | |||
BC095466.1 | - | 4 | 415 |