Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TIRAP SIRNA Assays ›
          Gene Symbol:
          TIRAP
          Gene Name:
          TIR domain containing adaptor protein
          Gene Aliases:
          BACTS1, Mal, MyD88-2, wyatt
          Chromosome Location:
          Chr.11: 126281268 - 126294933 on Build GRCh38
          UniGene ID:
          Hs.537126
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS150912
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TIRAP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039661.1 NP_001034750.1 4 469
          NM_001318776.1 NP_001305705.1 3 316
          NM_001318777.1 NP_001305706.1 3 316
          NM_148910.2 NP_683708.1 4 485
          XM_005271399.3 XP_005271456.2 2 214
          XM_011542576.2 XP_011540878.1 2 214
          XM_017017162.1 XP_016872651.1 4 506
          GenBank mRNA AB446477.1 - 1 33
          AF378129.1 - 4 466
          AF406652.1 - 1 33
          AF410783.1 - 4 462
          AK124298.1 - 3 316
          AK313147.1 - 3 309
          AY576785.1 - 1 72
          AY576786.1 - 1 72
          AY576787.1 - 1 72
          BC032474.1 - 4 438 5216171

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MyD88-dependent toll-like receptor signaling pathway
          inflammatory response
          cell surface receptor signaling pathway
          activation of NF-kappaB-inducing kinase activity
          myeloid cell differentiation
          positive regulation of B cell proliferation
          response to lipopolysaccharide
          regulation of interferon-beta production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-15 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of toll-like receptor 2 signaling pathway
          positive regulation of toll-like receptor 3 signaling pathway
          toll-like receptor 4 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          TIRAP-dependent toll-like receptor 4 signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of growth of symbiont in host
          innate immune response
          regulation of innate immune response
          positive regulation of interleukin-6 biosynthetic process
          positive regulation of JNK cascade
          defense response to Gram-positive bacterium
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of ERK1 and ERK2 cascade
          3'-UTR-mediated mRNA stabilization
          cellular response to bacterial lipopeptide
          cellular response to lipoteichoic acid
          positive regulation of neutrophil chemotaxis
          positive regulation of protein homodimerization activity
          positive regulation of chemokine (C-X-C motif) ligand 1 production
          positive regulation of chemokine (C-X-C motif) ligand 2 production
          protein kinase C binding
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          protein binding, bridging
          Toll-like receptor 4 binding
          Toll-like receptor 2 binding
          protein homodimerization activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0