Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CARD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017534.1 | NP_001017534.1 | 3 | 584 | ||
NM_052889.2 | NP_443121.1 | 4 | 647 | |||
XM_011542583.2 | XP_011540885.1 | 3 | 776 | |||
GenBank mRNA | AY885669.1 | - | 3 | 575 |