Search
Search

CCDC155| Gene Symbol: CCDC155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144688.4 | NP_653289.3 | 7 | 808 | ||
| XM_005258549.3 | XP_005258606.1 | 7 | 808 | |||
| XM_011526488.2 | XP_011524790.1 | 7 | 808 | |||
| XM_011526489.2 | XP_011524791.1 | 7 | 2708 | |||
| XM_011526490.2 | XP_011524792.1 | 7 | 808 | |||
| XM_011526494.2 | XP_011524796.1 | 8 | 781 | |||
| XM_011526495.2 | XP_011524797.1 | 8 | 817 | |||
| XM_011526496.2 | XP_011524798.1 | 6 | 669 | |||
| XM_011526497.2 | XP_011524799.1 | 7 | 767 | |||
| XM_017026333.1 | XP_016881822.1 | 6 | 941 | |||
| GenBank mRNA | AK057220.1 | - | 7 | 777 | ||
| AK097696.1 | - | 8 | 1521 | |||
| BC029811.1 | - | 7 | 727 | 5171712 | ||