Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182532.2 | NP_872338.1 | 3 | 934 | ||
XM_005270586.4 | XP_005270643.1 | 3 | 572 | |||
XM_011540911.2 | XP_011539213.1 | 4 | 686 | |||
XM_011540912.2 | XP_011539214.1 | 3 | 1462 | |||
GenBank mRNA | AA918832.1 | - | 1 | 284 | ||
BC029775.1 | - | 3 | 806 | 5186268 |