Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RBPMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194272.1 | NP_919248.1 | 5 | 604 | ||
GenBank mRNA | AK124123.1 | - | 5 | 352 | ||
AK127873.1 | - | 5 | 620 | |||
AY369207.1 | - | 5 | 604 | |||
BC131612.1 | - | 4 | 256 | |||
BC148208.1 | - | 4 | 256 |