Search
Search

| Gene Symbol: LINC00863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135292.1 | 5 | 2217 | |||
| GenBank mRNA | BC082979.1 | - | 5 | 2024 | ||
| CA439404.1 | - | 1 | 230 | |||
| DB313025.1 | - | 1 | 219 | |||
| Gene Symbol: NUTM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011540122.2 | XP_011538424.1 | 3 | 2256 | ||
| XM_017016607.1 | XP_016872096.1 | 3 | 2257 | |||