Search
Search

FAM179A| Gene Symbol: FAM179A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321538.1 | NP_001308467.1 | 4 | 633 | ||
| NM_001321539.1 | NP_001308468.1 | 4 | 633 | |||
| NM_199280.2 | NP_954974.2 | 4 | 633 | |||
| XM_011532628.2 | XP_011530930.1 | 4 | 633 | |||
| XM_017003507.1 | XP_016858996.1 | 4 | 682 | |||
| XM_017003512.1 | XP_016859001.1 | 4 | 633 | |||
| GenBank mRNA | AK296977.1 | - | 4 | 633 | ||
| BI768759.1 | - | 4 | 603 | |||