Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ACCSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031854.2 | NP_001027025.2 | 4 | 718 | ||
XM_017017714.1 | XP_016873203.1 | 7 | 1091 | |||
XM_017017715.1 | XP_016873204.1 | 4 | 731 | |||
GenBank mRNA | AB231732.1 | - | 4 | 297 | ||
AB231734.1 | - | 4 | 718 | |||
BC144554.1 | - | 4 | 731 |