Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZFP14 SIRNA Assays ›
          Gene Symbol:
          ZFP14 CR1 DPP9 LINC00562 LOC440864 MARCH2 MDM2 MDN1 MIPOL1 MRPL45P2 NQO1 ORAI2 OVOS PPIAL4C RNF103 SERPINB1 SLC40A1 TBRG4 TPM4 TSNAX TTC1 YIPF4 ZKSCAN1
          Gene Name:
          ZFP14 zinc finger protein
          complement component 3b/4b receptor 1 (Knops blood group)
          dipeptidyl peptidase 9
          long intergenic non-protein coding RNA 562
          uncharacterized LOC440864
          membrane associated ring-CH-type finger 2
          MDM2 proto-oncogene
          midasin AAA ATPase 1
          mirror-image polydactyly 1
          mitochondrial ribosomal protein L45 pseudogene 2
          NAD(P)H quinone dehydrogenase 1
          ORAI calcium release-activated calcium modulator 2
          ovostatin
          peptidylprolyl isomerase A like 4C
          ring finger protein 103
          serpin family B member 1
          solute carrier family 40 member 1
          transforming growth factor beta regulator 4
          tropomyosin 4
          translin associated factor X
          tetratricopeptide repeat domain 1
          Yip1 domain family member 4
          zinc finger with KRAB and SCAN domains 1
          Gene Aliases:
          ZNF531
          C3BR, C4BR, CD35, KN
          DP9, DPLP9, DPRP-2, DPRP2
          -
          -
          HSPC240, MARCH-II, RNF172
          ACTFS, HDMX, hdm2
          Rea1
          CCDC193
          -
          DHQU, DIA4, DTD, NMOR1, NMORI, QR1
          C7orf19, CBCIP2, MEM142B, TMEM142B
          -
          COAS-2
          HKF-1, KF-1, KF1, ZFP-103, ZFP103
          EI, ELANH2, HEL-S-27, HEL57, LEI, M/NEI, MNEI, PI-2, PI2
          FPN1, HFE4, IREG1, MST079, MSTP079, MTP1, SLC11A3
          CPR2, FASTKD4
          HEL-S-108
          TRAX
          TPR1
          FinGER4, Nbla11189
          9130423L19Rik, KOX18, PHZ-37, ZNF139, ZNF36, ZSCAN33
          Chromosome Location:
          Chr.19: 36334453 - 36379203 on Build GRCh38
          Chromosome Location:
          Chr.1: 207496128 - 207641765 on Build GRCh38
          Chromosome Location:
          Chr.19: 4675226 - 4723843 on Build GRCh38
          Chromosome Location:
          Chr.13: 47930153 - 47932622 on Build GRCh38
          Chromosome Location:
          Chr.2: 47695881 - 47708867 on Build GRCh38
          Chromosome Location:
          Chr.19: 8413272 - 8439021 on Build GRCh38
          Chromosome Location:
          Chr.12: 68808149 - 68845544 on Build GRCh38
          Chromosome Location:
          Chr.6: 89642775 - 89828021 on Build GRCh38
          Chromosome Location:
          Chr.14: 37197888 - 37579207 on Build GRCh38
          Chromosome Location:
          Chr.17: 47450227 - 47492620 on Build GRCh38
          Chromosome Location:
          Chr.16: 69709401 - 69726668 on Build GRCh38
          Chromosome Location:
          Chr.7: 102433530 - 102456821 on Build GRCh38
          Chromosome Location:
          Chr.12: 9517803 - 9573287 on Build GRCh38
          Chromosome Location:
          Chr.1: 149583823 - 149584607 on Build GRCh38
          Chromosome Location:
          Chr.2: 86603393 - 86623877 on Build GRCh38
          Chromosome Location:
          Chr.6: 2832332 - 2842049 on Build GRCh38
          Chromosome Location:
          Chr.2: 189560590 - 189580811 on Build GRCh38
          Chromosome Location:
          Chr.7: 45100100 - 45111747 on Build GRCh38
          Chromosome Location:
          Chr.19: 16067507 - 16103005 on Build GRCh38
          Chromosome Location:
          Chr.1: 231528653 - 231566524 on Build GRCh38
          Chromosome Location:
          Chr.5: 160009100 - 160065545 on Build GRCh38
          Chromosome Location:
          Chr.2: 32277889 - 32307929 on Build GRCh38
          Chromosome Location:
          Chr.7: 100015572 - 100045523 on Build GRCh38
          UniGene ID:
          Hs.35524
          Hs.334019
          Hs.515081
          Hs.660978
          Hs.388171
          Hs.631861
          Hs.484551
          Hs.529948
          Hs.660396
          Hs.537279
          Hs.406515
          Hs.363308
          -
          Hs.534674
          Hs.469199
          Hs.381167
          Hs.643005
          Hs.231411
          Hs.631618
          Hs.13318
          Hs.519718
          Hs.468099
          Hs.615360
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID HSS161289
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZFP14
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001297619.1 NP_001284548.1 5 5665
          NM_020917.2 NP_065968.1 5 5662
          XM_017027038.1 XP_016882527.1 5 5942
          GenBank mRNA AB046779.1 - 4 5635
          BC152446.1 - 4 5635 8862575
          Gene Symbol: CR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000573.3 NP_000564.2 39 8233
          NM_000651.4 NP_000642.3 47 9583
          XM_006711166.2 XP_006711229.1 46 9454
          XM_011509205.1 XP_011507507.1 48 9720
          GenBank mRNA BX113709.1 - 1 393
          Gene Symbol: DPP9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA CB960096.1 - 4 337
          Gene Symbol: LINC00562
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_047492.1 1 727
          Gene Symbol: LOC440864
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC040724.1 - 1 1080
          Gene Symbol: MARCH2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016496.4 NP_057580.3 2 147
          XM_017026853.1 XP_016882342.1 2 372
          GenBank mRNA AF151074.1 - 2 57
          BC032624.1 - 1 161 5241282
          BI546028.1 - 2 161
          Gene Symbol: MDM2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145337.2 NP_001138809.1 11 1745
          NM_001145339.2 NP_001138811.1 9 1971
          NM_001145340.2 NP_001138812.1 5 1298
          NM_001278462.1 NP_001265391.1 6 1376
          NM_002392.5 NP_002383.2 11 2136
          XM_005268872.4 XP_005268929.1 11 2064
          XM_006719399.3 XP_006719462.1 9 1719
          XM_006719400.3 XP_006719463.1 10 2025
          GenBank mRNA M92424.1 - 11 2122
          Z12020.1 - 11 2122
          Gene Symbol: MDN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK124025.1 - 10 1898
          Gene Symbol: MIPOL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011536463.2 XP_011534765.1 16 2059
          XM_017020998.1 XP_016876487.1 17 2166
          XM_017020999.1 XP_016876488.1 18 2242
          XM_017021000.1 XP_016876489.1 14 1726
          XM_017021001.1 XP_016876490.1 15 1865
          XM_017021002.1 XP_016876491.1 11 1316
          XM_017021019.1 XP_016876508.1 7 4631
          Gene Symbol: MRPL45P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC090855.1 - 4 403
          Gene Symbol: NQO1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000903.2 NP_000894.1 6 1682
          NM_001025433.1 NP_001020604.1 5 1580
          NM_001025434.1 NP_001020605.1 5 1568
          NM_001286137.1 NP_001273066.1 4 1504
          GenBank mRNA BM787983.1 - 1 314
          J03934.1 - 6 1540
          Gene Symbol: ORAI2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001126340.2 NP_001119812.1 4 5141
          NM_001271818.1 NP_001258747.1 4 5164
          NM_001271819.1 NP_001258748.1 3 4903
          NM_032831.3 NP_116220.1 3 5032
          Gene Symbol: OVOS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BX647938.1 - 5 3701
          Gene Symbol: PPIAL4C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK123006.1 - 2 3834
          Gene Symbol: RNF103
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK122978.1 - 1 125
          Gene Symbol: SERPINB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_030666.3 NP_109591.1 7 2210
          NR_073111.1 8 2342
          NR_073112.1 7 2206
          XM_011514333.1 XP_011512635.1 8 2433
          XM_011514334.2 XP_011512636.1 7 2344
          XM_011514335.2 XP_011512637.1 7 2199
          GenBank mRNA BM823453.1 - 1 381
          Gene Symbol: SLC40A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BX537503.1 - 1 523
          Gene Symbol: TBRG4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB023165.1 - 9 3095
          Gene Symbol: TPM4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011528232.1 XP_011526534.1 1 889
          XM_017027189.1 XP_016882678.1 1 889
          GenBank mRNA AK096732.1 - 1 889
          Gene Symbol: TSNAX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005999.2 NP_005990.1 6 1823
          GenBank mRNA BC010376.1 - 6 1781 4283594
          BC011797.1 - 6 1798 4138517
          BX537610.1 - 5 4791
          X95073.1 - 6 1825
          Gene Symbol: TTC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK128840.1 - 1 1844
          Gene Symbol: YIPF4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005264599.3 XP_005264656.1 6 1659
          Gene Symbol: ZKSCAN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK123012.1 - 1 721

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme chromatin/chromatin-binding, or -regulatory protein oxidoreductase ion channel transporter protease inhibitor actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          complement receptor mediated signaling pathway
          complement activation, classical pathway
          regulation of complement activation
          innate immune response
          negative regulation of complement activation, alternative pathway
          negative regulation of complement activation, classical pathway
          viral entry into host cell
          negative regulation of serine-type endopeptidase activity
          positive regulation of serine-type endopeptidase activity
          endocytosis
          protein ubiquitination
          negative regulation of transcription from RNA polymerase II promoter
          blood vessel development
          blood vessel remodeling
          regulation of heart rate
          atrioventricular valve morphogenesis
          endocardial cushion morphogenesis
          ventricular septum development
          atrial septum development
          protein complex assembly
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          traversing start control point of mitotic cell cycle
          positive regulation of cell proliferation
          response to toxic substance
          response to carbohydrate
          response to iron ion
          positive regulation of gene expression
          negative regulation of protein processing
          viral process
          protein sumoylation
          peptidyl-lysine modification
          protein destabilization
          response to magnesium ion
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          protein localization to nucleus
          regulation of protein catabolic process
          response to cocaine
          response to drug
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to morphine
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          establishment of protein localization
          response to ether
          negative regulation of transcription, DNA-templated
          positive regulation of mitotic cell cycle
          response to antibiotic
          positive regulation of protein export from nucleus
          response to steroid hormone
          cardiac septum morphogenesis
          cellular response to hydrogen peroxide
          negative regulation of cell cycle arrest
          cellular response to antibiotic
          cellular response to vitamin B1
          cellular response to alkaloid
          cellular response to growth factor stimulus
          cellular response to peptide hormone stimulus
          cellular response to estrogen stimulus
          cellular response to hypoxia
          cellular response to UV-C
          regulation of signal transduction by p53 class mediator
          negative regulation of signal transduction by p53 class mediator
          regulation of cellular amino acid metabolic process
          xenobiotic metabolic process
          nitric oxide biosynthetic process
          synaptic transmission, cholinergic
          aging
          response to nutrient
          removal of superoxide radicals
          response to estradiol
          negative regulation of catalytic activity
          positive regulation of neuron apoptotic process
          response to ethanol
          oxidation-reduction process
          store-operated calcium entry
          calcium ion transmembrane transport
          negative regulation of endopeptidase activity
          osteoblast differentiation
          movement of cell or subcellular component
          muscle contraction
          muscle filament sliding
          multicellular organism development
          spermatogenesis
          protein transport
          cell differentiation
          gene silencing by RNA
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding
          virus receptor activity
          complement component C3b binding
          complement component C4b binding
          complement component C4b receptor activity
          complement component C3b receptor activity
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          p53 binding
          SUMO transferase activity
          enzyme binding
          ubiquitin protein ligase binding
          identical protein binding
          peroxisome proliferator activated receptor binding
          ubiquitin protein ligase activity
          scaffold protein binding
          NAD(P)H dehydrogenase (quinone) activity
          cytochrome-b5 reductase activity, acting on NAD(P)H
          superoxide dismutase activity
          poly(A) RNA binding
          store-operated calcium channel activity
          serine-type endopeptidase inhibitor activity
          actin binding
          calcium ion binding
          structural constituent of muscle
          protein transporter activity
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0