Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MAGI2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038343.2 | 1 | 197 | |||
NR_038344.1 | 1 | 197 | ||||
GenBank mRNA | AK311103.1 | - | 1 | 165 | ||
BI521274.1 | - | 1 | 197 | |||
CN311982.1 | - | 1 | 164 |