Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNRNPA1 SIRNA Assays ›
          Gene Symbol:
          HNRNPA1 HNRNPA1L2 HNRNPA1P10
          Gene Name:
          heterogeneous nuclear ribonucleoprotein A1
          heterogeneous nuclear ribonucleoprotein A1-like 2
          heterogeneous nuclear ribonucleoprotein A1 pseudogene 10
          Gene Aliases:
          ALS19, ALS20, HNRPA1, HNRPA1L3, IBMPFD3, UP 1, hnRNP A1, hnRNP-A1
          -
          HNRPA1L-2
          Chromosome Location:
          Chr.12: 54280690 - 54287088 on Build GRCh38
          Chromosome Location:
          Chr.13: 52617470 - 52643784 on Build GRCh38
          Chromosome Location:
          Chr.19: 11665736 - 11667089 on Build GRCh38
          UniGene ID:
          Hs.546261
          Hs.447506
          Hs.534822
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID HSS162272
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HNRNPA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002136.3 NP_002127.1 10 1224
          NM_031157.3 NP_112420.1 11 1380
          XM_017019251.1 XP_016874740.1 9 1143
          GenBank mRNA AK129814.1 - 10 1166
          AK298176.1 - 7 724
          AK308679.1 - 8 1349
          BC002355.2 - 10 1145 2821751
          BC004945.1 - 1 1041
          BC009600.1 - 10 1167 4134638
          BC012158.1 - 10 1127 4564655
          BC020442.2 - 1 1134
          BC033714.1 - 10 1156 4420039
          BC035253.1 - 6 583
          BC052296.1 - 10 1136 6297642
          BC071945.1 - 1 1156 5475712
          BC073162.1 - 1 1150 6154972
          BC074502.1 - 10 1149 3455742
          BC103707.1 - 9 975 6154800
          BC121133.2 - 10 1150 40120894
          BU501222.1 - 1 597
          U00947.1 - 10 1114
          X04347.1 - 7 733
          X06747.1 - 10 1191
          Gene Symbol: HNRNPA1L2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001011724.1 NP_001011724.1 7 2179
          NM_001011725.1 NP_001011725.1 6 2038
          GenBank mRNA AK091082.1 - 1 1891
          AK094423.1 - 6 1929
          AK123159.1 - 5 1718
          AK126454.1 - 1 2813
          BC082242.1 - 6 1969
          BC108266.1 - 1 1167 4778431
          Gene Symbol: HNRNPA1P10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002944.2 1 1164

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          mRNA processing
          RNA export from nucleus
          fibroblast growth factor receptor signaling pathway
          gene expression
          viral process
          negative regulation of telomere maintenance via telomerase
          positive regulation of telomere maintenance via telomerase
          mRNA transport
          nuclear export
          nuclear import
          RNA splicing
          nucleotide binding
          single-stranded DNA binding
          RNA binding
          single-stranded RNA binding
          protein binding
          poly(A) RNA binding
          telomeric repeat-containing RNA binding
          G-rich strand telomeric DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline