Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: YBX3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027011.1 | 1 | 399 | |||
GenBank mRNA | BI826429.1 | - | 1 | 107 | ||
DB451883.1 | - | 1 | 370 | |||
DB459603.1 | - | 1 | 399 |