Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534891.2 | XP_011533193.1 | 14 | 3427 | ||
XM_011534893.2 | XP_011533195.1 | 13 | 3145 | |||
XM_017020365.1 | XP_016875854.1 | 14 | 3317 | |||
XM_017020366.1 | XP_016875855.1 | 14 | 3192 | |||
XM_017020367.1 | XP_016875856.1 | 14 | 3226 | |||
XM_017020368.1 | XP_016875857.1 | 14 | 3238 |
Gene Symbol: LINC00268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126863.1 | - | 2 | 2421 |