Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004051.4 | NP_004042.1 | 7 | 2998 | ||
NM_203314.2 | NP_976059.1 | 8 | 3150 | |||
NM_203315.2 | NP_976060.1 | 7 | 2934 | |||
XM_005269352.3 | XP_005269409.1 | 7 | 2814 | |||
XM_005269355.4 | XP_005269412.1 | 3 | 2767 | |||
XM_011513067.2 | XP_011511369.1 | 6 | 4363 | |||
XM_017007007.1 | XP_016862496.1 | 7 | 4442 | |||
XM_017007008.1 | XP_016862497.1 | 8 | 5062 | |||
XM_017007009.1 | XP_016862498.1 | 9 | 4017 | |||
XM_017007010.1 | XP_016862499.1 | 8 | 3865 | |||
XM_017007011.1 | XP_016862500.1 | 10 | 4075 | |||
XM_017007012.1 | XP_016862501.1 | 9 | 3923 | |||
XM_017007014.1 | XP_016862503.1 | 4 | 4039 | |||
XM_017007015.1 | XP_016862504.1 | 5 | 2795 | |||
GenBank mRNA | AK126701.1 | - | 4 | 3707 |