Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006248.3 | NP_006239.3 | 1 | 78 | ||
GenBank mRNA | BX484538.1 | - | 1 | 78 |
Gene Symbol: PRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005039.3 | NP_005030.2 | 1 | 80 | ||
NM_199353.2 | NP_955385.1 | 1 | 80 | |||
NM_199354.2 | NP_955386.1 | 1 | 80 | |||
GenBank mRNA | BC044827.1 | - | 1 | 66 | ||
BC141917.1 | - | 1 | 80 | 40113876 | ||
BX485626.1 | - | 1 | 79 | |||
BX487087.1 | - | 1 | 79 | |||
K03204.1 | - | 1 | 80 | |||
K03205.1 | - | 1 | 68 | |||
K03206.1 | - | 1 | 76 |