Search
Search

| Gene Symbol: PRAMEF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101343.2 | - | 4 | 1344 | 40025151 | |
| BC137391.1 | - | 4 | 1344 | 9021389 | ||
| BC171794.1 | - | 4 | 1344 | 9054448 | ||
| Gene Symbol: PRAMEF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC144426.1 | - | 4 | 1334 | 9052947 | |
| Gene Symbol: PRAMEF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098376.2 | NP_001091846.1 | 4 | 1358 | ||