Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POLR2J4 SIRNA Assays ›
          Gene Symbol:
          POLR2J4 SPDYE1 SPDYE18 SPDYE2 SPDYE2B SPDYE6
          Gene Name:
          RNA polymerase II subunit J4, pseudogene
          speedy/RINGO cell cycle regulator family member E1
          speedy/RINGO cell cycle regulator family member E18
          speedy/RINGO cell cycle regulator family member E2
          speedy/RINGO cell cycle regulator family member E2B
          speedy/RINGO cell cycle regulator family member E6
          Gene Aliases:
          RPB11-phi
          Ringo1, SPDYE, WBSCR19
          -
          SPDYB2-L1
          SPDYE2L
          -
          Chromosome Location:
          Chr.7: 43940895 - 44019194 on Build GRCh38
          Chromosome Location:
          Chr.7: 43997855 - 44011040 on Build GRCh38
          Chromosome Location:
          Chr.7: 77051689 - 77060537 on Build GRCh38
          Chromosome Location:
          Chr.7: 102551232 - 102565227 on Build GRCh38
          Chromosome Location:
          Chr.7: 102650325 - 102663525 on Build GRCh38
          Chromosome Location:
          Chr.7: 102345752 - 102356444 on Build GRCh38
          UniGene ID:
          Hs.657028
          Hs.645483
          -
          Hs.656428
          Hs.433879
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID HSS167797
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POLR2J4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC006435.1 - 1 359
          BC017341.2 - 4 477
          Gene Symbol: SPDYE1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005249719.3 XP_005249776.1 2 582
          XM_017012015.1 XP_016867504.1 2 520
          XM_017012017.1 XP_016867506.1 1 614
          Gene Symbol: SPDYE18
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005250742.4 XP_005250799.1 1 80
          XM_011516756.2 XP_011515058.1 1 80
          Gene Symbol: SPDYE2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001031618.3 NP_001026789.2 2 529
          XM_011516237.2 XP_011514539.1 2 528
          XM_017012223.1 XP_016867712.1 2 528
          GenBank mRNA AK302217.1 - 2 529
          JQ023160.1 - 1 55
          Gene Symbol: SPDYE2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166339.1 NP_001159811.1 2 529
          XM_005250093.3 XP_005250150.1 1 1097
          XM_011515702.2 XP_011514004.1 1 1097
          Gene Symbol: SPDYE6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146210.2 NP_001139682.2 1 476

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein kinase activity
          protein kinase binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0