Search Thermo Fisher Scientific
Gene Symbol: LOC100130920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093639.1 | - | 1 | 589 |
Gene Symbol: PRKCQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717465.3 | XP_006717528.1 | 1 | 589 | ||
XM_017016410.1 | XP_016871899.1 | 1 | 589 | |||
XM_017016411.1 | XP_016871900.1 | 1 | 589 |