Search
Search

| Gene Symbol: IGHM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X17115.1 | - | 2 | 279 | ||
| Gene Symbol: IGHG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC025314.1 | - | 2 | 252 | ||
| BC075846.1 | - | 2 | 249 | |||
| Gene Symbol: IGHV1-69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC070333.1 | - | 2 | 265 | ||