Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ARPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024959.2 | NP_001020130.1 | 5 | 690 | ||
NM_001024960.2 | NP_001020131.1 | 5 | 545 | |||
NM_001198780.1 | NP_001185709.1 | 5 | 544 | |||
NM_005718.4 | NP_005709.1 | 5 | 1056 | |||
GenBank mRNA | AF006087.1 | - | 5 | 497 | ||
AF019888.1 | - | 4 | 482 | |||
BC012596.2 | - | 4 | 479 | |||
BG938423.1 | - | 5 | 528 | |||
BI768875.1 | - | 5 | 670 | |||
BM045872.1 | - | 5 | 556 | |||
BQ688329.1 | - | 5 | 628 | |||
BX419672.2 | - | 5 | 544 |