Search
Search

ARPC3| Gene Symbol: ARPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278556.1 | NP_001265485.1 | 5, 6 | 536 | ||
| GenBank mRNA | AF004561.1 | - | 4, 5 | 385 | ||
| AF006086.1 | - | 5, 6 | 410 | |||
| AK297309.1 | - | 5, 6 | 467 | |||
| AK311789.1 | - | 5, 6 | 483 | |||
| BC067747.1 | - | 5, 6 | 442 | 5302338 | ||
| BC078162.1 | - | 5, 6 | 419 | 6148231 | ||
| BG723870.1 | - | 5, 6 | 500 | |||
| BM999380.1 | - | 2, 3 | 420 | |||
| CR407667.1 | - | 4, 5 | 385 | |||
| DQ328220.1 | - | 5, 6 | 466 | |||