Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI2 SIRNA Assays ›
Gene Symbol:
ABI2
Gene Name:
abl-interactor 2
Gene Aliases:
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
Chromosome Location:
Chr.2: 203327874 - 203432173 on Build GRCh38
UniGene ID:
Hs.471156
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS173427
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282925.1
NP_001269854.1
2
535
NM_001282926.1
NP_001269855.1
2
535
NM_001282927.1
NP_001269856.1
1
135
NM_001282932.1
NP_001269861.1
2
531
NM_005759.5
NP_005750.4
2
535
XM_005246217.2
XP_005246274.1
2
900
XM_005246218.2
XP_005246275.1
2
900
XM_005246221.2
XP_005246278.1
2
900
XM_005246225.2
XP_005246282.1
2
900
XM_005246226.2
XP_005246283.1
2
900
XM_006712174.2
XP_006712237.1
2
900
XM_006712177.2
XP_006712240.1
2
900
XM_006712179.2
XP_006712242.1
2
900
XM_006712180.2
XP_006712243.1
2
900
XM_006712181.2
XP_006712244.1
2
900
XM_006712182.2
XP_006712245.1
2
900
XM_006712185.1
XP_006712248.1
2
900
XM_006712186.2
XP_006712249.1
2
900
XM_011510472.2
XP_011508774.1
2
900
XM_011510473.2
XP_011508775.1
1
142
XM_017003118.1
XP_016858607.1
2
900
XM_017003119.1
XP_016858608.1
2
900
XM_017003120.1
XP_016858609.1
2
900
XM_017003121.1
XP_016858610.1
2
900
XM_017003122.1
XP_016858611.1
2
900
XM_017003123.1
XP_016858612.1
2
900
XM_017003124.1
XP_016858613.1
2
900
XM_017003125.1
XP_016858614.1
2
900
XM_017003126.1
XP_016858615.1
2
900
XM_017003127.1
XP_016858616.1
2
900
XM_017003128.1
XP_016858617.1
2
900
XM_017003129.1
XP_016858618.1
2
900
XM_017003130.1
XP_016858619.1
2
900
XM_017003131.1
XP_016858620.1
2
900
XM_017003132.1
XP_016858621.1
2
900
XM_017003133.1
XP_016858622.1
2
900
XM_017003134.1
XP_016858623.1
2
900
XM_017003135.1
XP_016858624.1
2
900
XM_017003136.1
XP_016858625.1
2
900
XM_017003137.1
XP_016858626.1
2
900
XM_017003138.1
XP_016858627.1
2
900
XM_017003139.1
XP_016858628.1
2
900
XM_017003140.1
XP_016858629.1
2
900
XM_017003141.1
XP_016858630.1
2
900
XM_017003142.1
XP_016858631.1
2
900
XM_017003143.1
XP_016858632.1
2
900
XM_017003144.1
XP_016858633.1
2
900
XM_017003145.1
XP_016858634.1
2
900
XM_017003146.1
XP_016858635.1
2
900
GenBank mRNA
AF260261.1
-
2
259
AK299824.1
-
2
379
AK302853.1
-
2
457
AK316000.1
-
2
365
BC001439.2
-
2
284
3138341
BT009920.1
-
2
259
DC376086.1
-
2
384
U23435.1
-
1
96
X95632.1
-
2
359
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
cytoskeleton organization
actin polymerization or depolymerization
viral process
cell migration
Rac protein signal transduction
peptidyl-tyrosine phosphorylation
positive regulation of Arp2/3 complex-mediated actin nucleation
DNA binding
protein binding
cytoskeletal adaptor activity
SH3 domain binding
kinase binding
ubiquitin protein ligase binding
protein complex binding
Rac GTPase binding
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0