Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABCA7 SIRNA Assays ›
          Gene Symbol:
          ABCA7
          Gene Name:
          ATP binding cassette subfamily A member 7
          Gene Aliases:
          ABCA-SSN, ABCX, AD9
          Chromosome Location:
          Chr.19: 1040103 - 1065572 on Build GRCh38
          UniGene ID:
          Hs.134514
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS173577
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABCA7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019112.3 NP_061985.2 14 2061
          XM_006722616.1 XP_006722679.1 14 2048
          XM_006722617.2 XP_006722680.1 14 2048
          XM_011527628.2 XP_011525930.1 14 1922
          XM_011527629.1 XP_011525931.1 14 2048
          XM_011527630.1 XP_011525932.1 14 2048
          XM_011527631.1 XP_011525933.1 14 2048
          XM_011527632.1 XP_011525934.1 10 1500
          XM_011527633.2 XP_011525935.1 14 2048
          XM_011527634.1 XP_011525936.1 14 2048
          XM_011527635.1 XP_011525937.1 14 2048
          XM_017026142.1 XP_016881631.1 14 2048
          XM_017026143.1 XP_016881632.1 14 2048
          GenBank mRNA AB055390.1 - 8 1416
          AF250238.1 - 13 1830
          AF328787.1 - 13 1946

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          phagocytosis
          memory
          positive regulation of cholesterol efflux
          phospholipid scrambling
          peptide cross-linking
          negative regulation of ATPase activity
          positive regulation of ATPase activity
          cholesterol efflux
          phospholipid efflux
          high-density lipoprotein particle assembly
          protein localization to nucleus
          apolipoprotein A-I-mediated signaling pathway
          negative regulation of amyloid precursor protein biosynthetic process
          positive regulation of phagocytosis
          transmembrane transport
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of beta-amyloid clearance
          positive regulation of engulfment of apoptotic cell
          negative regulation of beta-amyloid formation
          positive regulation of phospholipid efflux
          transporter activity
          ATP binding
          phospholipid transporter activity
          ATPase activity
          apolipoprotein A-I receptor activity
          ATPase activity, coupled to transmembrane movement of substances

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pqldv:80/100.66.72.197:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0