Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAP1 SIRNA Assays ›
Gene Symbol:
CAP1
Gene Name:
adenylate cyclase associated protein 1
Gene Aliases:
CAP, CAP1-PEN
Chromosome Location:
Chr.1: 40040070 - 40072649 on Build GRCh38
UniGene ID:
Hs.370581
Species:
Human
siRNA ID
HSS173675
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105530.1
NP_001099000.1
12
1529
NM_006367.3
NP_006358.1
12
1547
XM_005270368.1
XP_005270425.1
12
1387
XM_011540509.1
XP_011538811.1
13
1940
XM_011540510.1
XP_011538812.1
13
1937
XM_011540511.1
XP_011538813.1
12
1371
XM_011540512.1
XP_011538814.1
12
1490
XM_011540513.2
XP_011538815.1
12
1382
XM_011540514.1
XP_011538816.1
13
1497
XM_011540515.1
XP_011538817.1
13
1937
XM_017000067.1
XP_016855556.1
12
1455
XM_017000068.1
XP_016855557.1
13
1499
XM_017000069.1
XP_016855558.1
13
1710
XM_017000070.1
XP_016855559.1
12
1729
XM_017000071.1
XP_016855560.1
12
1687
XM_017000072.1
XP_016855561.1
12
1623
XM_017000073.1
XP_016855562.1
12
1444
GenBank mRNA
AK096193.1
-
12
1381
AK222513.1
-
12
1388
AK295396.1
-
13
1309
AK297829.1
-
12
1256
AK298094.1
-
12
1227
AK298110.1
-
12
1331
AK300861.1
-
11
1226
AK315731.1
-
12
1434
BC013963.2
-
12
1364
2963922
BC020367.1
-
1
86
BC095440.1
-
12
1369
30370256
BT007152.1
-
11
1318
CR457409.1
-
11
1318
L12168.1
-
11
1357
M98474.1
-
12
1388
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
ameboidal-type cell migration
receptor-mediated endocytosis
establishment or maintenance of cell polarity
signal transduction
activation of adenylate cyclase activity
actin polymerization or depolymerization
actin binding
adenylate cyclase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-bn9m5:80/10.239.167.51:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0