Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NEBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173484.1 | NP_001166955.1 | 7 | 1147 | ||
NM_006393.2 | NP_006384.1 | 28 | 3424 | |||
NM_213569.2 | NP_998734.1 | 7 | 1149 | |||
XM_005252342.4 | XP_005252399.1 | 27 | 3426 | |||
XM_005252343.4 | XP_005252400.1 | 26 | 3285 | |||
XM_005252344.4 | XP_005252401.1 | 27 | 3329 | |||
XM_011519291.2 | XP_011517593.1 | 30 | 3420 | |||
XM_017015467.1 | XP_016870956.1 | 30 | 3536 | |||
XM_017015468.1 | XP_016870957.1 | 31 | 3304 | |||
GenBank mRNA | AB209311.1 | - | 7 | 1021 | ||
AJ580772.1 | - | 7 | 1149 | |||
AK096540.1 | - | 7 | 771 | |||
AK294926.1 | - | 6 | 695 | |||
AK295186.1 | - | 7 | 804 | |||
AK316521.1 | - | 6 | 683 | |||
BC110452.1 | - | 28 | 3831 | 40027448 | ||
BC110453.1 | - | 28 | 3111 | 40027449 | ||
BC126132.1 | - | 7 | 1073 | |||
BC126134.1 | - | 7 | 1073 | |||
Y16241.1 | - | 28 | 3424 |