Search
Search

LBX1| Gene Symbol: LBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006562.4 | NP_006553.2 | 2 | 554 | ||
| GenBank mRNA | AJ004949.1 | - | 1 | 19 | ||
| BC069156.1 | - | 2 | 555 | 7217017 | ||
| BC136321.1 | - | 2 | 526 | 9020308 | ||
| X74863.1 | - | 1 | 13 | |||
| X90828.1 | - | 2 | 547 | |||