Search
Search

CETN3| Gene Symbol: CETN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297765.1 | NP_001284694.1 | 3, 4 | 393 | ||
| NM_001297768.1 | NP_001284697.1 | 3, 4 | 393 | |||
| NM_004365.3 | NP_004356.2 | 3, 4 | 393 | |||
| GenBank mRNA | BC005383.1 | - | 3, 4 | 363 | 3935674 | |
| BC093793.1 | - | 3, 4 | 329 | 7939638 | ||
| BC112040.1 | - | 3, 4 | 329 | 8327508 | ||
| BM553126.1 | - | 3, 4 | 373 | |||
| BT006688.1 | - | 3, 4 | 268 | |||
| BU569574.1 | - | 4, 5 | 342 | |||
| BU633961.1 | - | 2, 3 | 601 | |||
| CB988525.1 | - | 3, 4 | 415 | |||
| CB998764.1 | - | 3, 4 | 396 | |||
| CR450346.1 | - | 3, 4 | 268 | |||
| Y12473.1 | - | 4, 5 | 268 | |||