Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAPGEF4 SIRNA Assays ›
          Gene Symbol:
          RAPGEF4
          Gene Name:
          Rap guanine nucleotide exchange factor 4
          Gene Aliases:
          CAMP-GEFII, CGEF2, EPAC, EPAC 2, EPAC2, Nbla00496
          Chromosome Location:
          Chr.2: 172735282 - 173052893 on Build GRCh38
          UniGene ID:
          Hs.470646
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS174078
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAPGEF4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100397.1 NP_001093867.1 12 1341
          NM_001282899.1 NP_001269828.1 12 1212
          NM_001282900.1 NP_001269829.1 11 1158
          NM_001282901.1 NP_001269830.1 9 946
          NM_007023.3 NP_008954.2 15 1582
          XM_005246246.2 XP_005246303.1 14 1395
          XM_006712205.3 XP_006712268.1 14 1400
          XM_011510505.2 XP_011508807.1 14 1429
          XM_017003196.1 XP_016858685.1 15 1449
          XM_017003197.1 XP_016858686.1 11 1098
          GenBank mRNA AB027471.1 - 15 1452
          AB209681.1 - 15 3636
          AK294433.1 - 9 870
          AK294445.1 - 12 1061
          AK294914.1 - 9 899
          AK294997.1 - 14 1335
          AK295002.1 - 11 1093
          AK296340.1 - 11 1153
          AK302732.1 - 10 937
          AK304278.1 - 12 1101
          AK311165.1 - 11 980
          AK313084.1 - 15 1451
          AK316014.1 - 12 1176
          AK316072.1 - 12 1108
          BC024004.2 - 15 1421 4823935
          BC040183.2 - 12 1342 4801092
          KJ534924.1 - 15 1359
          KJ534925.1 - 14 1341
          KJ534978.1 - 15 1395
          U78516.1 - 15 1503

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_008761973, AB031229.
          This siRNA also targets the following transcripts in Mouse: NM_001204166, NM_019688, NM_001204165, AB021132, AK203721, and others.

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          small GTPase mediated signal transduction
          calcium ion regulated exocytosis
          regulation of exocytosis
          cAMP-mediated signaling
          insulin secretion
          positive regulation of GTPase activity
          regulation of insulin secretion
          guanyl-nucleotide exchange factor activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          Ras GTPase binding
          cAMP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0